Skip to Content
Merck
All Photos(5)

Key Documents

HPA012759

Sigma-Aldrich

Anti-NECTIN2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

CD112, HVEB, PRR2, PVRL2, PVRR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VRVQVLPEVRGQLGGTVELPCHLLPPVPGLYISLVTWQRPDAPANHQNVAAFHPKMGPSFPSPKPGSERLSFVSAKQSTGQDTEAELQDATLALHGLTVEDEGNYTCEFATFPKGSVRGMTWLR

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PVRL2(5819)

General description

Poliovirus receptor-related protein 2 (PVRL2) is a member of the immunoglobulin (Ig) superfamily. It contains three homologous Ig domains, an N-terminal variable-like (V) domain and two constant-like (C2) domains. PVRL2 is localized to the cadherin-based adherens junctions and cell junctions. The gene encoding this protein is present on chromosome 19q13.2-q13.4.

Immunogen

nectin cell adhesion molecule 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-PVRL2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Poliovirus receptor-related protein 2 (PVRL2) plays an important role as a glycoprotein D (gD) receptor for the entry of variant forms of herpes simplex virus 1 (HSV-1), herpes simplex virus 2 (HSV-2) and porcine pseudorabies virus (PRV). PVRL2 also has a role in cell-to-cell adhesion. It has been shown that knockout of the PVRL2 gene in mice results in male sterility because of the defects in spermatogenesis. PVRL2 is a ligand for DNAX Accessory Molecule-1 (DNAM-1) activating receptor. It′s binding and interaction enhances the natural killer (NK)-mediated lysis of tumor cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71916

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Whelan et al.
Cancer immunology research, 7(2), 257-268 (2019-01-20)
Although checkpoint inhibitors that block CTLA-4 and PD-1 have improved cancer immunotherapies, targeting additional checkpoint receptors may be required to broaden patient response to immunotherapy. PVRIG is a coinhibitory receptor of the DNAM/TIGIT/CD96 nectin family that binds to PVRL2. We
F Eberlé et al.
Gene, 159(2), 267-272 (1995-07-04)
Until now it was assumed that the murine poliovirus (PV) receptor homolog gene (MPH) had been identified. Alternative splicing of MPH transcripts generates two glycoproteins named MPH alpha and MPH beta which share an identical N-terminal region composed of three
Structural features of nectin-2 (HveB) required for herpes simplex virus entry.
WM Martinez and PG Spear
Journal of Virology, 75(22), 11185-11195 (2001)
Daniela Pende et al.
Molecular immunology, 42(4), 463-469 (2004-12-21)
The capability of NK lymphocytes to kill tumor cells depends on different receptors/ligands interactions. In order to identify the cellular ligands recognized by "orphan" triggering receptors, mice were immunized with NK susceptible target cells. mAbs were selected that inhibited NK
Daniel Wai-Hung Ho et al.
Nature communications, 12(1), 3684-3684 (2021-06-19)
Interaction between tumor cells and immune cells in the tumor microenvironment is important in cancer development. Immune cells interact with the tumor cells to shape this process. Here, we use single-cell RNA sequencing analysis to delineate the immune landscape and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service