Key Documents
EP5804735005
Eppendorf® Fixed-angle rotor
to hold, 6 x 8-tube or 5-tube PCR strips, to hold, 48 x 0.2 mL PCR tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 48 x 0.2 mL PCR tubes
to hold 6 x 8-tube or 5-tube PCR strips
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5804735005
parameter
12,000 max. RCF (15,294 xg)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG