EP0030000650
Eppendorf® epT.I.P.S. standard
volume × L 0.2-5.0 mL × 175 mm , L, violet, pack of 300 ea (3 bags × 100 tips)
Synonym(s):
pipetting tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
maximum volume:
5 mL
volume × L:
0.2-5.0 mL × 175 mm , L
sterility:
non-sterile
feature:
Eppendorf Quality™
packaging:
pack of 300 ea (3 bags × 100 tips)
manufacturer/tradename:
Eppendorf® 0030000650
Recommended Products
material
violet
size
L
sterility
non-sterile
feature
Eppendorf Quality™
packaging
pack of 300 ea (3 bags × 100 tips)
manufacturer/tradename
Eppendorf® 0030000650
maximum volume
5 mL
volume × L
0.2-5.0 mL × 175 mm , L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Bulk packed in re-sealable bags.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
Eppendorf® epT.I.P., are optimally coordinated to Eppendorf pipettes and, meets the EN ISO 8655 requirements. This results in minimal attachment and ejection forces with the highest level of tightness.
Application
- Pipetting liquids
- Dispensing liquids
- Mixing liquids
- Filling plates and reaction vessels
- Filling gels for electrophoresis
- Phase extraction and removal of supernatants
Features and Benefits
- Original, high-quality Eppendorf pipette tips packaged in resealable bags, epT.I.P.S.
- Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes.
- Graduations for easy visualization of pipetting volumes.
- Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service