Skip to Content
Merck
All Photos(1)

Key Documents

EMU010011

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Tlr2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGACACTGGGGGTAACATCGCTTTTTCCCAATCTCACAAATTTACAAACCCTCAGGATAGGAAATGTAGAGACTTTCAGTGAGATAAGGAGAATAGATTTTGCTGGGCTGACTTCTCTCAATGAACTTGAAATTAAGGCATTAAGTCTCCGGAATTATCAGTCCCAAAGTCTAAAGTCGATCCGCGACATCCATCACCTGACTCTTCACTTAAGCGAGTCTGCTTTCCTGCTGGAGATTTTTGCAGATATTCTGAGTTCTGTGAGATATTTAGAACTAAGAGATACTAACTTGGCCAGGTTCCAGTTTTCACCACTGCCCGTAGATGAAGTCAGCTCACCGATGAAGAAGCTGGCATTCCGAGGCTCGGTTCTCACTGATGAAAGCTTTAACGAGCTCCTGAAGCTGTTGCGTTACA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung Heon Shin et al.
Allergy, asthma & immunology research, 8(1), 63-68 (2015-11-06)
Chronic rhinosinusitis with nasal polyps is a chronic inflammatory disease with markedly increased eosinophils, Th2-type lymphocytes, fibroblasts, and goblet cells. Fungi are commonly associated with airway inflammatory diseases, and thymic stromal lymphopoietin (TSLP) is important in the development of Th2
Min Li et al.
Biochemical and biophysical research communications, 466(4), 748-754 (2015-10-02)
Microphage apoptosis is a critical event in atherosclerotic lesions in patients with diabetes. In the present investigation, high glucose treatment inhibited Akt phosphorylation and activated caspase 3 in primary peritoneal macrophage, leading to cell apoptosis. Hypoxia prolonged macrophage survival in
Megumi Inomata et al.
PloS one, 13(8), e0202791-e0202791 (2018-08-29)
Porphyromonas gingivalis possesses various abilities to evade and disrupt host immune responses, by which it acts as an important periodontal pathogen. P. gingivalis produces outer membrane protein A (OmpA)-like proteins (OmpALPs), Pgm6 and Pgm7, as major O-linked glycoproteins, but their
Seok-Seong Kang et al.
Cytokine, 75(1), 174-180 (2015-05-20)
Staphylococcus aureus can cause the intestinal inflammatory diseases. However, little is known about the molecular mechanism of S. aureus infection in the intestine. In the present study, we investigated whether S. aureus could stimulate human intestinal epithelial cells triggering inflammation.
Helge Haarmann et al.
Biochemical and biophysical research communications, 467(1), 46-52 (2015-09-30)
Bacterial colonisation with Moraxella catarrhalis may partly sustain chronic inflammation in the lower airways of patients with chronic obstructive pulmonary disease (COPD). In addition, this bacterium causes infectious exacerbations of COPD, which often necessitate treatment with antibiotics. Antimicrobial peptides are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service