Skip to Content
Merck
All Photos(1)

Key Documents

EHU100071

Sigma-Aldrich

MISSION® esiRNA

targeting human NUAK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCATCACCACAAGCACAACTTGAAGCACCGCTACGAGCTGCAGGAGACCCTGGGCAAAGGCACCTACGGCAAAGTCAAGCGGGCCACCGAGAGGTTTTCTGGCCGAGTGGTTGCTATAAAATCCATTCGTAAGGACAAAATTAAGGATGAACAAGACATGGTTCACATCAGACGAGAGATTGAGATCATGTCATCTCTCAACCATCCTCATATCATCAGTATTTATGAAGTGTTTGAGAACAAAGATAAGATTGTGATCATCATGGAATATGCCAGCAAAGGGGAGCTGTACGATTACATCAGTGAGCGGCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Kui Peng et al.
Oncology letters, 15(3), 3639-3645 (2018-02-23)
Hypoxia is a common characteristic of solid tumors. Previous studies have reported that the tumor invasion-associated factor, AMPK-related protein kinase 5 (ARK5), is associated with a poor prognosis in colon cancer. However, whether or not ARK5 is involved in hypoxia
Minghui Li et al.
American journal of translational research, 9(4), 1708-1719 (2017-05-05)
Lung cancer incidence and mortality rates are amongst the highest of all malignant tumors worldwide. ARK5 is a member of the human AMP-activated protein kinase (AMPK) family which is implicated in tumor survival and progression. The current study was designed
Emilia Escalona et al.
Frontiers in oncology, 10, 1123-1123 (2020-08-06)
NUAK1 is an AMPK-related kinase located in the cytosol and the nucleus, whose expression associates with tumor malignancy and poor patient prognosis in several cancers. Accordingly, NUAK1 was associated with metastasis because it promotes cell migration and invasion in different
Martin Golkowski et al.
Cell systems, 11(2), 196-207 (2020-08-07)
Hepatocellular carcinoma (HCC) is a complex and deadly disease lacking druggable genetic mutations. The limited efficacy of systemic treatments for advanced HCC implies that predictive biomarkers and drug targets are urgently needed. Most HCC drugs target protein kinases, indicating that
Giacomo Cossa et al.
Molecular cell, 77(6), 1322-1339 (2020-02-02)
Deregulated expression of MYC induces a dependence on the NUAK1 kinase, but the molecular mechanisms underlying this dependence have not been fully clarified. Here, we show that NUAK1 is a predominantly nuclear protein that associates with a network of nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service