Skip to Content
Merck
All Photos(1)

Key Documents

EHU067421

Sigma-Aldrich

MISSION® esiRNA

targeting human LRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCTACTGCAACAGCAGCTTTCAGCTTCAGGCAGATGGCAAGACCTGCAAAGATTTTGATGAGTGCTCAGTGTACGGCACCTGCAGCCAGCTATGCACCAACACAGACGGCTCCTTCATATGTGGCTGTGTTGAAGGATACCTCCTGCAGCCGGATAACCGCTCCTGCAAGGCCAAGAACGAGCCAGTAGACCGGCCCCCTGTGCTGTTGATAGCCAACTCCCAGAACATCTTGGCCACGTACCTGAGTGGGGCCCAGGTGTCTACCATCACACCTACGAGCACGCGGCAGACCACAGCCATGGACTTCAGCTATGCCAACGAGACCGTATGCTGGGTGCATGTTGGGGACAGTGCTGCTCAGACGCAGCTCAAGTGTGCCCGCATGCCTGGCCTAAAGGGCTTCGTGGATGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianhua Peng et al.
Redox biology, 21, 101121-101121 (2019-02-01)
White matter injury (WMI) is associated with motor deficits and cognitive dysfunctions in subarachnoid hemorrhage (SAH) patients. Therapeutic strategy targeting WMI would likely improve the neurological outcomes after SAH. Low-density lipoprotein receptor-related protein-1 (LRP1), a scavenger receptor of apolipoprotein E
Tomoya Kitakaze et al.
Biochimica et biophysica acta. Molecular cell research, 1867(2), 118563-118563 (2019-11-02)
Skeletal muscle secretes biologically active proteins that contribute to muscle hypertrophy in response to either exercise or dietary intake. The identification of skeletal muscle-secreted proteins that induces hypertrophy can provide critical information regarding skeletal muscle health. Dietary provitamin A, β-carotene
Paola Merino et al.
The Journal of biological chemistry, 292(7), 2741-2753 (2016-12-18)
Axonal injury is a common cause of neurological dysfunction. Unfortunately, in contrast to axons from the peripheral nervous system, the limited capacity of regeneration of central nervous system (CNS) axons is a major obstacle for functional recovery in patients suffering
Ling Lin et al.
Oncotarget, 8(50), 88094-88103 (2017-11-21)
Macrophage accumulation is one of the hallmarks of progressive kidney disease. In response to injury, macrophages undergo a phenotypic polarization to become two functionally distinct subsets: M1 and M2 macrophages. Macrophage polarization is a dynamic process, and recent work indicates
Aline Appert-Collin et al.
Cell adhesion & migration, 11(4), 316-326 (2016-07-28)
The low-density lipoprotein receptor-related protein-1 (LRP-1) is a member of Low Density Lipoprotein Receptor (LDLR) family, which is ubiquitously expressed and which is described as a multifunctional endocytic receptor which mediates the clearance of various extracellular matrix molecules including serine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service