Skip to Content
Merck
All Photos(1)

Key Documents

EHU009271

Sigma-Aldrich

MISSION® esiRNA

targeting human PPM1D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGTCAGAGCTGTGGAGGTGACACAGGACCATAAGCCAGAACTTCCCAAGGAAAGAGAACGAATCGAAGGACTTGGTGGGAGTGTAATGAACAAGTCTGGGGTGAATCGTGTAGTTTGGAAACGACCTCGACTCACTCACAATGGACCTGTTAGAAGGAGCACAGTTATTGACCAGATTCCTTTTCTGGCAGTAGCAAGAGCACTTGGTGATTTGTGGAGCTATGATTTCTTCAGTGGTGAATTTGTGGTGTCACCTGAACCAGACACAAGTGTCCACACTCTTGACCCTCAGAAGCACAAGTATATTATATTGGGGAGTGATGGACTTTGGAATATGATTCCACCACAAGATGCCATCTCAATGTGCCAGGACCAAGAGGAGAAAAAATACCTGATGGGTGAGCATGGACAATCTTGTGCCAAAATGCTTGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Chen et al.
Journal of Cancer, 11(11), 3216-3224 (2020-04-02)
Accumulated studies showed that numerous microRNAs (miRNAs) were aberrantly expressed in human intrahepatic cholangiocarcinoma (ICC) and contributed to the tumorigenic processes. However, whether miR-129-2-3p is implicated in the ICC initiation and progression is still limited. Here, the results revealed that
Zhong-Wu Lu et al.
Oncology reports, 43(3), 783-794 (2020-01-11)
Endeavors towards identifying key molecular markers for early diagnosis and treatment are driving the clinical study of papillary thyroid carcinoma (PTC). Recent studies have indicated that protein phosphatase, Mg2+/Mn2+ dependent, 1D (PPM1D) exerts an oncogenic function by increasing cell proliferation, migration
Dong-Seok Park et al.
EMBO reports, 21(5), e48693-e48693 (2020-02-28)
The tumor suppressor Smad4, a key mediator of the TGF-β/BMP pathways, is essential for development and tissue homeostasis. Phosphorylation of Smad4 in its linker region catalyzed by the mitogen-activated protein kinase (MAPK) plays a pivotal role in regulating its transcriptional
Shigeo Ohba et al.
Cell reports, 31(2), 107518-107518 (2020-04-16)
The metabolic enzyme phosphoglycerate mutase 1 (PGAM1) is overexpressed in several types of cancer, suggesting an additional function beyond its established role in the glycolytic pathway. We here report that PGAM1 is overexpressed in gliomas where it increases the efficiency
Jin Ju Park et al.
Technology in cancer research & treatment, 19, 1533033820964425-1533033820964425 (2020-10-24)
Several techniques have been employed for deletion of the NKX3.1 gene, resulting in developmental defects of the prostate, including alterations in ductal branching morphogenesis and prostatic secretions as well as epithelial hyperplasia and dysplasia. To investigate whether the CRISPR/Cas9-mediated technique

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service