Skip to Content
Merck
All Photos(1)

Key Documents

EHU003851

Sigma-Aldrich

MISSION® esiRNA

targeting human SAMHD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACGCATGAACAAGGCTCAGTTATGATGTTTGAGCACCTTATTAATTCTAATGGAATTAAGCCTGTCATGGAACAATATGGTCTCATCCCTGAAGAAGATATTTGCTTTATAAAGGAACAAATTGTAGGACCACTTGAATCACCTGTCGAAGATTCATTGTGGCCATATAAAGGGCGTCCTGAAAACAAAAGCTTCCTTTATGAGATAGTATCTAATAAAAGAAATGGCATTGATGTGGACAAATGGGATTATTTTGCCAGGGACTGCCATCATCTTGGAATCCAAAATAATTTTGATTACAAGCGCTTTATTAAGTTTGCCCGTGTCTGTGAAGTAGACAATGAGTTGCGTATTTGTGCTAGAGATAAGGAAGTTGGAAATCTGTATGACATGTTCCACACTCGCAACTCTTTACACCGTAGAGCTTATCAACACAAAGTTGGCAACATTATTGATACAATGATTACAGATGCTTTCCTCAAAGCAGATGACTACATAGAGATTACAGGTGCTGGAGGAAAAAAGTATCGCATTTCTACAGCAATTGACGACATGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weihui Fu et al.
Scientific reports, 6, 38162-38162 (2016-12-07)
SAMHD1 restricts human immunodeficiency virus type 1 (HIV-1) replication in myeloid cells and CD4+ T cells, while Vpx can mediate SAMHD1 degradation to promote HIV-1 replication. Although the restriction mechanisms of SAMHD1 have been well-described, SAMHD1 expression and Vpx-mediated SAMHD1
Thomas Oellerich et al.
Nature communications, 10(1), 3475-3475 (2019-08-04)
Hypomethylating agents decitabine and azacytidine are regarded as interchangeable in the treatment of acute myeloid leukemia (AML). However, their mechanisms of action remain incompletely understood, and predictive biomarkers for HMA efficacy are lacking. Here, we show that the bioactive metabolite
Petra Mlcochova et al.
Cell reports, 30(12), 3972-3980 (2020-03-27)
Macrophages exist predominantly in two distinct states, G0 and a G1-like state that is accompanied by phosphorylation of SAMHD1 at T592. Here, we demonstrate that Toll-like receptor 4 (TLR4) activation can potently induce G0 arrest and SAMHD1 antiretroviral activity by
Simone De Meo et al.
PLoS pathogens, 16(9), e1008855-e1008855 (2020-09-29)
SAMHD1 is a host restriction factor that functions to restrict both retroviruses and DNA viruses, based on its nuclear deoxynucleotide triphosphate (dNTP) hydrolase activity that limits availability of intracellular dNTP pools. In the present study, we demonstrate that SAMHD1 expression
Vera Rocha-Perugini et al.
Nature microbiology, 2(11), 1513-1522 (2017-09-06)
In this study, we report that the tetraspanin CD81 enhances human immunodeficiency virus (HIV)-1 reverse transcription in HIV-1-infected cells. This is enabled by the direct interaction of CD81 with the deoxynucleoside triphosphate phosphohydrolase SAMHD1. This interaction prevents endosomal accumulation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service