Key Documents
CLS6796420D
Corning® hotplate and stirrer with digital display
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 5 in. × 7 in., CE compliant
Synonym(s):
stirrers
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
Pricing and availability is not currently available.
feature
CE compliant
packaging
case of 1
manufacturer/tradename
Corning 6796-420D
availability
not available in USA and Canada
parameter
5-550 °C temp. range
60-1150 rpm speed
plate L × W
5 in. × 7 in.
General description
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Microprocessor maintains consistent and repeatable temperature and stir speed settings.
  • Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
  • Separate temperature sensor provides power cut off if unit overheats
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)