Key Documents
CLS431355
Corning® foam centrifuge rack, 15 mL
Synonym(s):
Corning centrifuge
About This Item
UNSPSC Code:
41122804
NACRES:
NB.14
material
foamed PE
feature
Holds 15 mL tubes
packaging
pack of 20 ea
manufacturer/tradename
Corning 431355
availability
available only in South Korea
General description
Corning® expandable foam centrifuge rack holds 50 conical bottom plastic centrifuge tubes of 15 mL each.
Legal Information
Corning is a registered trademark of Corning, Inc.