Key Documents
CLS1300P500W
Corning® wide mouth wash bottle, reusable
capacity 500 mL, low-density polyethylene (LDPE), with GL-45 PP screw cap
Synonym(s):
laboratory wash bottles, plastic wash bottles
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
Pricing and availability is not currently available.
material
cylindrical bottle
low-density polyethylene (LDPE)
screw top cap
white bottle
wide-mouth bottle
packaging
pack of 12 ea
manufacturer/tradename
Corning 1300P-500W
H × O.D.
146 mm × 58 mm
capacity
500 mL
General description
  • Corning® reusable bottles are manufactured from low-density polyethylene (LDPE).
  • Features a wide mouth for easy transferring of samples.
  • Polypropylene cap and dispensing tube optimize backflow of the solution.
  • Unique design allows minimal turbulence of liquid in the bottle.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)