SPC212
11120717, human lung, Epitheloid/mesenchymal
Synonym(s):
SPC 212, SPC-212
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
SPC212, 11120717
biological source
human lung
growth mode
Adherent
karyotype
Not specified
morphology
Epitheloid/mesenchymal
receptors
Mesothelial-membrane antigen ME1
relevant disease(s)
cancer
Looking for similar products? Visit Product Comparison Guide
Cell Line Origin
A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure
Cell Line Description
SPC212 was derived from a biphasic tumour of the pleural cavity of a 47-year old female patient, prior to treatment, with a known history of exposure to asbestos.
DNA Profile
STR-PCR Data:
Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 9
D5S818: 11,12
D7S820: 8,10
THO1: 6,9
TPOX: 8
vWA: 16
Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 9
D5S818: 11,12
D7S820: 8,10
THO1: 6,9
TPOX: 8
vWA: 16
Culture Medium
Subculture Routine
Split subconfluent cultures (70-80%) 1:4 to 1:10 using 0.25% trypsin/EDTA; 5% CO2; 37 °C. Suggested seeding density 1-4 x 10,000 cells/cm2.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service