AE17 cell line from mouse lung
10092310, mouse lung, Cells are epithelial-like
Synonym(s):
AE17 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Product Name
AE17 cell line from mouse lung, 10092310
biological source
mouse lung
description
Mouse malignant mesothelioma
growth mode
Adherent
karyotype
Not specified
morphology
Cells are epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse malignant mesothelioma
Cell Line Description
Mice (C57Black/6J strain female 6-8 weeks old) were exposed to crocidolite asbestos through intraperitoneal injection resulting in tumour development (both ascites and solid tumours). Cultures were established from malignant mesothelial cells obtained from ascites fluid. Cells are tumourigenic in syngeneic immunocompetent mice. Cell Bank Australia reference number: CBA0156
Application
Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods.
Culture Medium
RPMI1640 (with 2mM L-Glutamine+25mM HEPES) + 5% FCS
Subculture Routine
Optimal split ratio 1:16 - 1:32 (seeding density 0.6 x104 cells/cm2).Cells incubated at 37°C with 5% CO2.Harvest the cells using 0.05% Trypsin/EDTA at 37°C for 5 min.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Richard Lake - University of Western Australia, Australia
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service