C9357
Cyclo(Ala-Arg-Gly-Asp-3-Aminomethylbenzoyl)
≥96% (HPLC), suitable for western blotting and enzyme immunoassays
Synonym(s):
XJ735
About This Item
Recommended Products
Product Name
Cyclo(Ala-Arg-Gly-Asp-3-Aminomethylbenzoyl), ≥96% (HPLC)
Quality Level
Assay
≥96% (HPLC)
form
powder
technique(s)
enzyme immunoassay: suitable
western blot: suitable
color
white
UniProt accession no.
storage temp.
−20°C
SMILES string
C[C@@H]1NC(=O)c2cccc(CNC(=O)[C@H](CC(O)=O)NC(=O)CNC(=O)[C@H](CCCNC(N)=N)NC1=O)c2
InChI
1S/C23H32N8O7/c1-12-19(35)31-15(6-3-7-26-23(24)25)21(37)28-11-17(32)30-16(9-18(33)34)22(38)27-10-13-4-2-5-14(8-13)20(36)29-12/h2,4-5,8,12,15-16H,3,6-7,9-11H2,1H3,(H,27,38)(H,28,37)(H,29,36)(H,30,32)(H,31,35)(H,33,34)(H4,24,25,26)/t12-,15-,16-/m0/s1
InChI key
HRASXYOWQQTBAQ-RCBQFDQVSA-N
Gene Information
human ... NPY(4852)
Related Categories
Amino Acid Sequence
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service