BAF187451300
Bel-Art® Switch-Grid® Replacement Grids
for tubes, 10 ‑ 13 mm, orange
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
material
orange
polyoxymethylene
feature
autoclavable
for tubes 10 ‑ 13 mm
manufacturer/tradename
Bel-Art F18745-1300
packaging
pack of 4 ×
Looking for similar products? Visit Product Comparison Guide
General description
1 RACK FRAME, 25 POSSIBLE TUBE SIZE COMBINATIONS
This innovative, split-frame rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Each rack frame includes a set of grids in one size. Grid sets in alternate sizes can be purchased as needed.
This innovative, split-frame rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Each rack frame includes a set of grids in one size. Grid sets in alternate sizes can be purchased as needed.
Features and Benefits
More compact and less expensive than purchasing multiple racks
Colored grid inserts correspond to a specific test tube size ranging from 10mm to 30mm in diameter
Three tier rack holds tubes securely in place
Rubber feet keep racks from sliding on benchtop, provide secure stacking while in storage
Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol or isopropyl alcohol; autoclaved at 121°C (250°F).
Colored grid inserts correspond to a specific test tube size ranging from 10mm to 30mm in diameter
Three tier rack holds tubes securely in place
Rubber feet keep racks from sliding on benchtop, provide secure stacking while in storage
Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol or isopropyl alcohol; autoclaved at 121°C (250°F).
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Switch-Grid is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service