AXYSCOLPB
Looped screw caps
with O-rings, blue, pkg of 8x500caps/cs
Synonym(s):
Corning screw cap
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Axygen Inc SCO-LP-B
Corning
packaging
pkg of 8x500caps/cs
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service