AXYHTF1000CBKHTRS
Axygen CO-RE style liquid level sensing filter tip
capacity 1000 μL, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
sterility
γ-irradiated
sterile
feature
liquid level sensing
packaging
case of 1536 tips racked
manufacturer/tradename
Corning HTF1000CBKHTRS
capacity
1000 μL
color
black
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service