AXYEV50RS
Pipette tips for Tecan® EVO/ Caliper Zephyr® Automated Liquid Handling System
racked, capacity 50 μL, pkg of 5x960tips/cs, clear, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
capacity:
50 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc EV-50-R-S
Corning
Corning
sterility
sterile
manufacturer/tradename
Axygen Inc EV-50-R-S
Corning
packaging
pkg of 5x960tips/cs
capacity
50 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Browse all pipette tips
Legal Information
Tecan is a registered trademark of Tecan Group AG
Zephyr is a registered trademark of Caliper Life Sciences, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service