Skip to Content
Merck
All Photos(1)

Key Documents

Y0001541

Risedronate sodium 2.5-hydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Risedronic acid monosodium salt hemi(pentahydrate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10NNaO7P2 · 2.5H2O
CAS Number:
Molecular Weight:
350.13
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

risedronate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OP(C(P(O)([O-])=O)(O)CC1=CC=CN=C1)(O)=O.[Na+]

InChI

1S/2C7H11NO7P2.2Na.5H2O/c2*9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6;;;;;;;/h2*1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15);;;5*1H2/q;;2*+1;;;;;/p-2

InChI key

HYFDYHPNTXOPPO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Risedronate sodium is a member of the pyridinyl class of bisphosphonates. It is mostly used as an antiresorptive agent. It can be used in treating and preventing postmenopausal and glucocorticoid-induced osteoporosis.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Risedronate sodium 2.5-hydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - STOT SE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service