Skip to Content
Merck
All Photos(1)

Key Documents

T5536

Supelco

Testosterone-d3 solution

100 μg/mL in 1,2-dimethoxyethane, analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

drug control

Home Office Schedule 4.2

concentration

100 μg/mL in 1,2-dimethoxyethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

[H]C1([H])C[C@]2([H])[C@@]3([H])CCC4=CC(=O)CC[C@]4(C)[C@]3([H])CC[C@@]2(C)[C@]1([H])O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1/i6D2,17D

InChI key

MUMGGOZAMZWBJJ-HZRGXLBSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service