NIST2586
Trace elements in soil containing lead from paint
NIST® SRM® 2586, nominal 500 mg/kg lead
Synonym(s):
Soil Trace Elements with Lead from Paint
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
Quality Level
packaging
pkg of 55 g
manufacturer/tradename
NIST®
application(s)
environmental
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Sources, Pathways, and Concentrations of Potentially Toxic Trace Metals in Home Environments: This study provides a detailed examination of trace elements, including lead from paint, in residential settings. It highlights the significant health risks associated with long-term exposure to trace metals in soils around homes, essential for regulatory actions and public health advisories (Ibañez-Del Rivero et al., 2023).
- Potential Health Risks of Lead Exposure: Focusing on the lifelong risks associated with lead exposure, particularly from contaminated soils, this research underscores the necessity for enhanced public health education to mitigate exposure risks from environments affected by lead-based paints (Olufemi et al., 2022).
- Lead Poisoning of Backyard Chickens: Implications for Urban Gardening and Food Production: Investigates how lead contamination in soil, often sourced from paint, affects backyard chickens, presenting a risk to urban agriculture. The study highlights the importance of assessing and managing soil quality to ensure the safety of homegrown food products (Yazdanparast et al., 2022).
- Lead in the Marine Environment: Concentrations and Effects on Invertebrates: Though focusing on marine settings, this review draws parallels with terrestrial ecosystems by discussing the toxicological impacts of lead on biological systems, relevant for understanding the broader ecological consequences of lead contamination from sources like paint (Botté et al., 2022).
- International Analysis of Sources and Human Health Risk Associated with Trace Metal Contaminants in Residential Indoor Dust: Offers a global perspective on the sources of trace metals in indoor environments, including lead from paint. This extensive analysis aids in developing strategies for reducing health risks associated with trace element exposure in residential and occupational settings (Isley et al., 2022).
Other Notes
Certified for the analytes listed below.
Arsenic (As), Cadmium (Cd), Chromium (Cr), Lead (Pb)
Reference/Informational value is provided for the following analytes.
Aluminum (Al), Neodymium (Nd), Barium (Ba), Phosphorus (P), Calcium (Ca), Potassium (K), Cerium (Ce), Praseodymium (Pr), Dysprosium (Dy), Samarium (Sm), Europium (Eu), Silicon (Si), Gadolinium (Gd), Sodium (Na), Iron (Fe), Strontium (Sr), Lanthanum (La), Terbium (Tb), Magnesium (Mg), Titanium (Ti), Manganese (Mn), Ytterbium (Yb), Mercury (Hg), Yttrium (Y), Zinc (Zn), Perfluorooctane Sulfonate (PFOS), Beryllium (Be), Nickel (Ni), Cobalt (Co), Niobium (Nb), Copper (Cu), Scandium (Sc), Erbium (Er), Selenium (Se), Gallium (Ga), Thorium (Th), Holmium (Ho), Thulium (Tm), Lithium (Li), Vanadium (V)
See certificate for values and more details at nist.gov/SRM.
Arsenic (As), Cadmium (Cd), Chromium (Cr), Lead (Pb)
Reference/Informational value is provided for the following analytes.
Aluminum (Al), Neodymium (Nd), Barium (Ba), Phosphorus (P), Calcium (Ca), Potassium (K), Cerium (Ce), Praseodymium (Pr), Dysprosium (Dy), Samarium (Sm), Europium (Eu), Silicon (Si), Gadolinium (Gd), Sodium (Na), Iron (Fe), Strontium (Sr), Lanthanum (La), Terbium (Tb), Magnesium (Mg), Titanium (Ti), Manganese (Mn), Ytterbium (Yb), Mercury (Hg), Yttrium (Y), Zinc (Zn), Perfluorooctane Sulfonate (PFOS), Beryllium (Be), Nickel (Ni), Cobalt (Co), Niobium (Nb), Copper (Cu), Scandium (Sc), Erbium (Er), Selenium (Se), Gallium (Ga), Thorium (Th), Holmium (Ho), Thulium (Tm), Lithium (Li), Vanadium (V)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service