Skip to Content
Merck
All Photos(1)

Key Documents

93753

Millipore

Davis Minimal Broth

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Minimal Broth, Davis

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

ammonium sulfate, 1 g/L
dextrose, 1 g/L
dipotassium phosphate, 7 g/L
magnesium sulfate, 0.1 g/L
monopotassium phosphate, 2 g/L
sodium citrate, 0.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

7.0±0.2 (25 °C)

application(s)

food and beverages
life science and biopharma

microbiology

suitability

nonselective for Escherichia coli
nonselective for coliforms

Application

Davis Minimal Broth is recommended for the isolation and characterization of nutrional mutants of Escherichia coli.

Preparation Note

Suspend 11.6 g in 1000 ml of purified water. Heat with frequent agitation and boil for one minute. Sterilize at 121°C for 15 minutes. Mix well.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service