Skip to Content
Merck
All Photos(1)

Key Documents

84135

Supelco

Micro particles based on polystyrene

analytical standard, size: 30 μm

Synonym(s):

Polystyrene latex particles (30 μm), Polystyrene microbeads (30 μm), Polystyrene microspheres (30 μm), Polystyrene microparticles (30 μm), Latex beads from PS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

form

aqueous suspension
particles

crosslinking

2 % cross-linked

concentration

10% (solids)

particle size

30 μm std dev <0.4 μm, coeff var <1%

density

1.05 g/cm3

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene based microparticles (size: 30 μm) are a particle size standard, ideal to determine particle size distribution (PSD) of test samples.

Application

Used for equipment particle size monitoring and validation.
Also used to:
  • investigate the impact of polystyrene microplastics on marine biota and organic matter
  • study optical confinement in optofluidic laser measurements

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • available in 5 and 10 mL pack sizes as neat samples

Analysis Note

For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast and reproducible ELISA laser platform for ultrasensitive protein quantification
Tan X, et al.
ACS sensors, 5(1) (2019)
Mild toxicity of polystyrene and polymethylmethacrylate microplastics in Paracentrotus lividus early life stages
Thomas PJ, et al.
Marine Environmental Research, 161, 105132-105132 (2020)
Luisa Galgani et al.
Scientific reports, 8(1), 14635-14635 (2018-10-04)
About 5 trillion plastic particles are present in our oceans, from the macro to the micro size. Like any other aquatic particulate, plastics and microplastics can create a micro-environment, within which microbial and chemical conditions differ significantly from the surrounding
Manel Essaidi-Laziosi et al.
The Journal of allergy and clinical immunology, 141(6), 2074-2084 (2017-08-12)
The leading cause of acute illnesses, respiratory viruses, typically cause self-limited diseases, although severe complications can occur in fragile patients. Rhinoviruses (RVs), respiratory enteroviruses (EVs), influenza virus, respiratory syncytial viruses (RSVs), and coronaviruses are highly prevalent respiratory pathogens, but because
Bernadett Boda et al.
Antiviral research, 156, 72-79 (2018-06-12)
Respiratory viral infections cause mild to severe diseases, such as common cold, bronchiolitis and pneumonia and are associated with substantial burden for society. To test new molecules for shortening, alleviating the diseases or to develop new therapies, relevant human in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service