Skip to Content
Merck
All Photos(2)

Key Documents

06030

Sigma-Aldrich

2-Propene-1-thiol

technical, ~60% (GC)

Synonym(s):

Allyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2SH
CAS Number:
Molecular Weight:
74.14
Beilstein:
1697523
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

form

liquid

concentration

~60% (GC)

refractive index

n20/D 1.4765 (lit.)

bp

67-68 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

functional group

thiol

storage temp.

−20°C

SMILES string

SCC=C

InChI

1S/C3H6S/c1-2-3-4/h2,4H,1,3H2

InChI key

ULIKDJVNUXNQHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

4.0 °F - closed cup

Flash Point(C)

-15.56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeşeren Saylan et al.
Sensors (Basel, Switzerland), 18(9) (2018-09-13)
Hemoglobin is an iron carrying protein in erythrocytes and also an essential element to transfer oxygen from the lungs to the tissues. Abnormalities in hemoglobin concentration are closely correlated with health status and many diseases, including thalassemia, anemia, leukemia, heart

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service