Skip to Content
Merck
All Photos(2)

Key Documents

VSWP01300

Millipore

MCE Membrane Filter, 0.025 μm Pore Size

MF-Millipore, filter diam. 13 mm, hydrophilic

Synonym(s):

Cellulose nitrate membrane filter discs, Hydrophilic MCE membrane filter discs, MF-Millipore Membrane Filter, 0.025 µm pore size, Mixed Cellulose Ester membrane filter discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

description

13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

0.15 L/min-cm2 air flow rate
0.15 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

13 mm

thickness

100 μm

gravimetric extractables

1.5%

refractive index

n/D 1.5

matrix

MF-Millipore

pore size

0.025 μm pore size
72 % porosity

bubble point

≥21.1 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Filter Code: VSWP

Application

Microdialysis of DNA and proteins

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service