MIACFD101
Microfil® Funnel Dispenser
For use with 100mL Microfil® funnels
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41116129
NACRES:
NB.24
Quality Level
packaging
pkg of 1 unit
manufacturer/tradename
Microfil® with S-Pak®
input
sample type food(s)
sample type beverage(s)
application(s)
bioburden testing
clinical
environmental
food and beverages
industrial qc
compatibility
for use with EZ-Fit® Filtration Manifold
shipped in
ambient
Related Categories
General description
The funnel dispenser for the 100 mL funnel is an additional accessory of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. The funnels, packed in convenient packs of 25, are loaded to the custom-designed funnel dispenser and can be easily removed as needed. The funnel dispenser is designed to allow rapid dispensing of Microfil® funnels without compromising sterility.
Application
The funnel dispenser is an additional accessory of the Microfil® filtration system used in general microbiological analysis of beer, bottled water, pharmaceutical products analysis, environmental analysis, bioburden testing, and water monitoring.
Legal Information
EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service