M000037P0
Fluid Contamination Monitor, empty
Reload cassette with appropriate filters
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24
material
Tenite™ plastic
polystyrene
Quality Level
sterility
non-sterile
filter diam.
37 mm
filtration area
9 cm2
fitting
female Luer inlet slip
female Luer outlet slip
application(s)
industrial qc
shipped in
ambient
General description
Fluid Contamination Analysis Monitors are transparent, disposable filter holders made from Tenite plastic and pre-assembled with MF-Millipore filters (effective filtration area is 9 cm2). A thick cellulose pad, 34 mm diameter, between the monitor sections seals the filter to distribute sample flow evenly over the filter surface. Use with Fluid Sampling Kit (XX6403730).
Matched-Weight Monitors:
Matched-Weight Monitors eliminate test filter pre-weighing in gravimetric analysis. Each monitor contains two filters matched in weight to within 0.1 mg. Sample passes through both, but upper (test) filter retains all contaminant. The difference in filter weights, after drying, equals the weight of contaminant in sample.
Matched-Weight Monitors:
Matched-Weight Monitors eliminate test filter pre-weighing in gravimetric analysis. Each monitor contains two filters matched in weight to within 0.1 mg. Sample passes through both, but upper (test) filter retains all contaminant. The difference in filter weights, after drying, equals the weight of contaminant in sample.
Principle
Vacuum
Legal Information
Tenite is a trademark of Eastman Chemical Company
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service