Skip to Content
Merck
All Photos(2)

Key Documents

ABN1680

Sigma-Aldrich

Anti-phospho-Pannexin 1 (Tyr308)

from rabbit

Synonym(s):

Panx-1, Panx1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on high sequence homology), chimpanzee (based on high sequence homology), rat (based on high sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pTyr308)

Gene Information

human ... PANX1(24145)

General description

Pannexin-1 (UniProt: P60570) is encoded by the Panx1 (also known as Px1) gene (Gene ID: 315435) in rat. Pannexin-1 is a multi-pass membrane protein of the pannexin family that serves as a high-conductance, voltage-gated channel. It is widely expressed, including in cartilage, skin, spleen, heart, and brain. In the nervous system Panx1 transcripts have been detected in the cerebellum, cortex, lens, retina, pyramidal cells, interneurons of the neocortex and hippocampus. Three pannexin family genes (PANX1-3) have been identified in murine and human genomes. Pannexins share structural and functional similarities and play important roles in several physiological processes, including ATP release, Ca2+ homeostasis, and synaptic neurotransmitters release. Pannexins can also form intermixed channels and the intermixing is more evident between Pannexin 1 and 2. The level of intermixing is governed by the degree of N-glycosylation. Post-translational N-glycosylation plays a role in cell surface targeting. Pannexin-1 can also undergo S-nitrosylation, which can inhibit channel currents and ATP release. It has been shown that Pannexin-1 channel can release ATP in response to activation by TNF-alpha that requires activation of type-1 TNF receptors and downstream signaling through Src family kinases (SFK). It is shown that NMDARs, Src kinase and Pannexin-1 form a signaling complex, and activation of Pannexin-1 requires phosphorylation at Tyr308. Excitotoxic NMDA and glycine stimulation of N2a cells expressing Pannexin-1 is shown to increase phosphorylation at Tyr308, but not in cells expressing Pannexin-1 with Y308F substitution. Pannexin-1 has been linked to propagation of calcium waves, regulation of vascular tone, taste-bud function, and in tumor suppression in gliomas. Pannexin-1 channel opening can contribute to cell death and seizures under ischemic or epileptic conditions. (Ref.: Penuela, S et al. (2013). Biochim. Biophys. Acta 1828(11); 15-22; Weilinger, N et al. (2016). Nat. Neurosci. 19(3): 432-442).

Specificity

This polyclonal antibody specifically detects pannexin 1 phosphorylated on tyrosine 308 in rat. Also predicted to react with human and chimpanzee.

Immunogen

A linear peptide corresponding to 13 amino acids surrounding phosphotyrosine 308 of rat pannexin-1.
Epitope: cytoplasmic domain

Application

Anti-phospho-Pannexin 1 (Tyr308) Antibody, Cat. No. ABN1680, is a highly specific rabbit polyclonal antibody that targets Pannexin 1 and has been tested in Immunohistochemistry and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected phospho-Pannexin 1 (Tyr308) in wt/mutant transfected N2a cells +/- stimulus (Weilinger, N.L., et. al. (2016). Nat Neurosci. 19(3):432-42).

Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-Pannexin 1 (Tyr308) in acute isolated hippocampal brain tissue slice (Courtesy of Dr Roger J. Thompson at University of Calgary).

Quality

Evaluated by Western Blotting in HEK293T cells transfected with WT rat Panx1 and dominant active Src kinase (Y530F Src).

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-Pannexin 1 (Tyr308) in 10 µg of HEK293T cells transfected with WT rat Panx1 and dominant active Src kinase (Y530F Src).

Target description

~55 kDa observed; 48.07 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS and 50% glycerol without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Ruan et al.
eLife, 13 (2024-05-23)
Protein phosphorylation is one of the major molecular mechanisms regulating protein activity and function throughout the cell. Pannexin 1 (PANX1) is a large-pore channel permeable to ATP and other cellular metabolites. Its tyrosine phosphorylation and subsequent activation have been found
Leon J DeLalio et al.
The Journal of biological chemistry, 294(17), 6940-6956 (2019-03-01)
Pannexin 1 (PANX1)-mediated ATP release in vascular smooth muscle coordinates α1-adrenergic receptor (α1-AR) vasoconstriction and blood pressure homeostasis. We recently identified amino acids 198-200 (YLK) on the PANX1 intracellular loop that are critical for α1-AR-mediated vasoconstriction and PANX1 channel function.
Lisa Maria Metz et al.
International journal of molecular sciences, 23(9) (2022-05-15)
Pannexin-1 (PANX1) is a transmembrane protein that forms ion channels as hexamers on the plasma membrane. Electrophysiological studies prove that PANX1 has a high conductance for adenosine triphosphate (ATP), which plays an important role as a signal molecule in platelet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service