941862
Dip tube
for use with 25 L combi container for withdrawl system (Cat No. 1.01114)
Synonym(s):
Dip tube 25 l combi for 1.01114.0001
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122409
NACRES:
NB.51
Recommended Products
compatibility
for use with 25 L combi container for withdrawl system (Cat No. 1.01114)
shipped in
ambient
storage temp.
room temp
Related Categories
General description
To customize the withdrawal systems for every requirement, we provide customers with the appropriate dip tubes for each package size suitable for the various withdrawal systems.
for 25 l combi container for withdrawal system Cat. No. 1.01114.0001
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service