Skip to Content
Merck
All Photos(1)

Key Documents

8.24566

Sigma-Aldrich

Platinum(II) chloride

73% Pt, for synthesis

Synonym(s):

Platinum(II) chloride (73% Pt), Platinum dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Pt
CAS Number:
Molecular Weight:
265.98
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-034-1
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

Platinum(II) chloride (73% Pt), for synthesis

Quality Level

form

powder

potency

3423 mg/kg LD50, oral (Rat)

pH

5-6 ( in H2O, suspension)

mp

581 °C (decomposes)

density

6.1 g/cm3

storage temp.

2-30°C

SMILES string

[Pt](Cl)Cl

InChI

1S/2ClH.Pt/h2*1H;/q;;+2/p-2

InChI key

CLSUSRZJUQMOHH-UHFFFAOYSA-L

Analysis Note

Assay (of Pt): ≥ 70.0 %

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service