Skip to Content
Merck
All Photos(1)

Key Documents

8.20524

Sigma-Aldrich

N,N-Diphenylacetamide

for synthesis

Synonym(s):

N,N-Diphenylacetamide, Acetic acid diphenylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(C6H5)2
CAS Number:
Molecular Weight:
211.26
MDL number:
UNSPSC Code:
12352111
EC Index Number:
208-277-1
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

powder

mp

100-103 °C (External MSDS)

storage temp.

2-30°C

SMILES string

N(c2ccccc2)(c1ccccc1)C(=O)C

InChI

1S/C14H13NO/c1-12(16)15(13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-11H,1H3

InChI key

DKLYDESVXZKCFI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service