Skip to Content
Merck
All Photos(2)

Key Documents

8.16021

Sigma-Aldrich

(S)-(-)-Tyrosine

for synthesis

Synonym(s):

(S)-(-)-Tyrosine, 4-Hydroxyphenylalanine, Tyr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO3
CAS Number:
Molecular Weight:
181.19
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-460-4
NACRES:
NA.22

Quality Level

form

powder

potency

>5110 mg/kg LD50, oral (Rat)

pH

6.5 (0.1 g/L in H2O)

mp

297-298 °C (decomposition)

solubility

0.38 g/L

density

1.46 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]C(Cc1ccc(cc1)O)C(=O)[O-]

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)

InChI key

OUYCCCASQSFEME-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=5 in HCl 1mol/l): -12.5 - -11.0 °
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service