693021
Zwittergent® 3-10 Detergent
A synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range.
Synonym(s):
n-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
About This Item
Recommended Products
Quality Level
Assay
≥99% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
aggregation number
41
color
white
CMC
25 - 40 mM
solubility
water: soluble
shipped in
ambient
storage temp.
15-25°C
SMILES string
[S](=O)(=O)([O-])CCC[N+](CCCCCCCCCC)(C)C
InChI
1S/C15H33NO3S/c1-4-5-6-7-8-9-10-11-13-16(2,3)14-12-15-20(17,18)19/h4-15H2,1-3H3
InChI key
WKALLSVICJPZTM-UHFFFAOYSA-N
Related Categories
General description
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service