Skip to Content
Merck
All Photos(1)

Key Documents

567550

Millipore

Sodium phosphate dibasic

ACS Reagent Grade. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers.

Synonym(s):

Sodium phosphate dibasic, Na₂HPO₄, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.25

grade

ACS reagent

Quality Level

description

Merck USA index - 14, 8659

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

impurities

≤0.01% insoluble material

color

white

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

water: soluble

cation traces

heavy metals: ≤0.001%

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

ACS Reagent Grade. Has a pH of 8.7-9.3.
ACS Reagent Grade. Useful in conjunction with Sodium Phosphate, Monobasic (Cat. No. 567545 or 567549) in the preparation of biological buffers.

Application


  • 5-Fluorouracil monodispersed chitosan microspheres: Microfluidic chip fabrication with crosslinking, characterization, drug release and anticancer activity.: This study utilizes sodium phosphate dibasic in the crosslinking process to create monodispersed chitosan microspheres for drug delivery. The microspheres are designed for the controlled release of 5-fluorouracil, showcasing potential applications in cancer therapy (He T et al., 2020).

  • Chitosan-based electrospun membranes: Effects of solution viscosity, coagulant and crosslinker.: The research investigates the use of sodium phosphate dibasic as a crosslinker in the fabrication of chitosan-based electrospun membranes. The findings highlight its significance in improving the structural properties of membranes for biomedical applications (Dodero A et al., 2020).

  • Sodium Phosphate.: This review provides a comprehensive overview of the applications and properties of sodium phosphate dibasic in various fields, including its roles in buffering solutions, food additives, and pharmaceutical formulations (NBK501909, 2006).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service