Skip to Content
Merck
All Photos(1)

Key Documents

5.04462

Sigma-Aldrich

IWR-1

InSolution, ≥95%, 25 mM in DMSO, Wnt Antagonist I, endo

Synonym(s):

InSolution Wnt Antagonist I, IWR-1-endo, Inhibitor of Wnt Response-1, TNKS1/2 Inhibitor II, Tankyrase1/2 Inhibitor II, Wnt Pathway Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19N3O3
CAS Number:
Molecular Weight:
409.44
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C25H19N3O3/c29-23(27-19-5-1-3-14-4-2-12-26-22(14)19)15-8-10-18(11-9-15)28-24(30)20-16-6-7-17(13-16)21(20)25(28)31/h1-12,16-17,20-21H,13H2,(H,27,29)/t16-,17+,20-,21+

InChI key

ZGSXEXBYLJIOGF-ALFLXDJESA-N

General description

A cell-permeable p-imidobenzamidoquinoline, endo-diastereomer that is shown to inhibit the activity of TNKS1/PARP5a and TNKS2/PARP5b in in vitro auto-PARsylation assays (IC50 = 131 and 56 nM, respectively) and effectively suppress Wnt-stimulated transcription activity in L-Wnt-STF-based reporter assays (IC50 = 180 nM), while exhibiting little activity against PARP1 or PARP2 (IC50 >18.75 µM). Although both IWR-1-endo and XAV939 (Tankyrase1/2 Inhibitor; >Cat. No. 575545) act as reversible Wnt pathway inhibitors and exhibit similar pharmacological effects both in vitro and in vivo, IWR-1-endo exerts its effect via interaction with Axin, while XAV939 binds TNKS directly.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Axin
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 25 mM (5 mg/489 µL) solution of Wnt Antagonist I, IWR-1-endo (Cat. No. 681669) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Wang, H., et al. 2011. ACS Chem. Biol.6, 192.
Chen, B., et al. 2009. Nature Chem. Biol.5, 100.
Huang, S.M., et al. 2009. Nature461, 614.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service