Skip to Content
Merck
All Photos(1)

Key Documents

288500-M

Millipore

DL-α-Difluoromethylornithine, Hydrochloride

A cell-permeable, anticancer agent that acts as a specific and irreversible inhibitor of ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine biosynthesis.

Synonym(s):

DL-α-Difluoromethylornithine, Hydrochloride, DFMO, Eflornithine, RMI-71782

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12F2N2O2 · xHCl · yH2O
CAS Number:
Molecular Weight:
182.17 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

description

Merck USA index - 14, 3522

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL
methanol: 25 mg/mL

storage temp.

10-30°C

SMILES string

FC(F)C(N)(CCCN)C(=O)O.[Cl-].O.[H+]

InChI

1S/C6H12F2N2O2.ClH.H2O/c7-4(8)6(10,5(11)12)2-1-3-9;;/h4H,1-3,9-10H2,(H,11,12);1H;1H2

InChI key

FJPAMFNRCFEGSD-UHFFFAOYSA-N

General description

A cell-permeable, anticancer agent that acts as a specific and irreversible inhibitor of ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine biosynthesis. Inhibits B16 melanoma-induced angiogenesis in ovo and proliferation of vascular endothelial cells in vitro.

Biochem/physiol Actions

Primary Target
ODC

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Meyskens, F.L., Jr., and Gerner, E.W. 1995. J. Cell. Biochem. (Suppl.) 22, 126.
Feuerstein, B.G., et al. 1992. Cancer Res. 52, 6782.
Takigawa, M., et al. 1990. Cancer Res.50, 4131.
Weeks, C.E., et al. 1982. Proc. Natl. Acad. Sci. USA 79, 6028.
Danzin, C., et al. 1979. Life Sci. 24, 519.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service