Skip to Content
Merck
All Photos(1)

Key Documents

230900

Sigma-Aldrich

Chymotrypsin, Human Pancreas

Natiave chymotrypsin from human pancreas. Pancreatic digestive enzyme. Increased levels of this enzyme and/or its zymogen have been found in serum of individuals with cystic fibrosis.

Synonym(s):

Chymotrypsin, Human Pancreas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human pancreas

Quality Level

description

Merck USA index - 14, 2260

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

ice cold 1 mM HCl: soluble

foreign activity

Trypsin, none detected (when assayed against N-benzyl-L-arginine-p-nitroanilide)

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

Natiave chymotrypsin from human pancreas. Pancreatic digestive enzyme. Increased levels of this enzyme and/or its zymogen have been found in serum of individuals with cystic fibrosis.

Warning

Toxicity: Harmful (C)

Physical form

Salt free.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Once opened, re-desiccate the dried product over silica gel under a vacuum and freeze (-20°C). Stock solutions are stable for up to 1 week at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service