Skip to Content
Merck
All Photos(1)

Key Documents

208904

Sigma-Aldrich

Calpastatin Peptide, Negative Control

A scrambled peptide with an identical amino acid composition to that of Calpastatin Peptide. Useful as a negative control for Calpastatin Peptide.

Synonym(s):

Calpastatin Peptide, Negative Control

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C142H230N36O44S
Molecular Weight:
3177.63
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 20 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

General description

A scrambled peptide with an identical amino acid composition to that of Calpastatin Peptide (Cat. No. 208902). Useful as a negative control for Calpastatin Peptide.
A scrambled peptide with an identical amino acid composition to that of Calpastatin Peptide (Cat. No. 208902). Useful as a negative control for Calpastatin Peptide.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Useful as a negative control for Calpastatin Peptide
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Ala-Pro-Arg-Leu-Glu-Ile-Val-Pro-Thr-Met-Tyr-Ile-Tyr-Lys-Leu-Ser-Pro-Thr-Gly-Ser-Glu-Lys-Leu-Glu-Asp-Glu-Arg-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -2-°C.

Other Notes

Eto, A., et al. 1995. J. Biol. Chem. 270, 25115.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service