Skip to Content
Merck
All Photos(1)

Key Documents

12-305

Sigma-Aldrich

3T3/A31 Cell Lysate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105330
eCl@ss:
32011807
NACRES:
NA.41

biological source

mouse

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

shipped in

dry ice

Cell Line Description

3T3/A31

Quality

Routinely evaluated as a positive control for some of Upstates antibodies.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexey V Karpushev et al.
Hypertension (Dallas, Tex. : 1979), 57(5), 996-1002 (2011-04-06)
The epithelial Na(+) channel (ENaC) is an essential channel responsible for Na(+) reabsorption in the aldosterone-sensitive distal nephron. Consequently, ENaC is a major effector impacting systemic blood volume and pressure. We have shown recently that Rac1 increases ENaC activity, whereas
Jason A Lehman et al.
The Journal of biological chemistry, 278(30), 28130-28138 (2003-05-13)
We report here for the first time the detection of the ribosomal p70S6 kinase (p70S6K) in a hematopoietic cell, the neutrophil, and the stimulation of its enzymatic activity by granulocyte macrophage colony-stimulating factor (GM-CSF). GM-CSF modified the Vmax of the
Sudhakaran Prabakaran et al.
Molecular systems biology, 7, 482-482 (2011-04-14)
The functional impact of multisite protein phosphorylation can depend on both the numbers and the positions of phosphorylated sites-the global pattern of phosphorylation or 'phospho-form'-giving biological systems profound capabilities for dynamic information processing. A central problem in quantitative systems biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service