Skip to Content
Merck
All Photos(1)

Key Documents

1.09011

Supelco

Nessler′s reagent A

for determination of nitrogen Solution A : potassium tetraiodomercurate(II) solution

Synonym(s):

Potassium mercury(II) iodide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NB.21

Quality Level

form

liquid

specific analyte(s)

nitrogen

technique(s)

photometry: suitable

pH

9.2 (20 °C in H2O)

density

1.25 g/cm3 at 20 °C

storage temp.

2-30°C

General description

for determination of nitrogen Solution A : potassium tetraiodomercurate(II) solution

Analysis Note

Application testconforms

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service