Skip to Content
Merck
All Photos(1)

Key Documents

1.06609

Supelco

Sodium peroxidisulfate

for analysis EMSURE®

Synonym(s):

Sodium peroxidisulfate, Sodium persulfate, Peroxydisulfuric acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O8S2
CAS Number:
Molecular Weight:
238.10
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-892-1
NACRES:
NA.21
Assay:
≥99.0% (iodometric)
form:
solid
solubility:
545 g/L
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥99.0% (iodometric)

form

solid

potency

920 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

loss

≤0.05% loss on drying, 105 °C; 3 hours

pH

3.5-3.8 (20 °C, 100 g/L in H2O)

solubility

545 g/L

density

1.2 g/cm3 at 20 °C

bulk density

1150 kg/m3

anion traces

chloride (Cl-): ≤0.01%

cation traces

Cr: ≤0.001%
Cu: ≤0.001%
Fe: ≤0.0005%
Mn: ≤0.0001%
NH4+: ≤0.05%
Pb: ≤0.001%
heavy metals (as Pb): ≤0.003%

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])OO[S](=O)(=O)[O-]

InChI

1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

CHQMHPLRPQMAMX-UHFFFAOYSA-L

Related Categories

Linkage

Replaces: SX0703; SX0703-1

Analysis Note

Assay (iodometric): ≥ 99.0 %
Identity: passes test
Chloride (Cl): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.003 %
NH₄ (Ammonium): ≤ 0.05 %
Cr (Chromium): ≤ 0.001 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Mn (Manganese): ≤ 0.0001 %
Pb (Lead): ≤ 0.001 %
Loss on drying (105 °C; 3 hours): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service