Skip to Content
Merck
All Photos(1)

Key Documents

1.06560

Supelco

Sodium perborate tetrahydrate

pure

Synonym(s):

Sodium perborate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BNaO3 · 4H2O
CAS Number:
Molecular Weight:
153.86
MDL number:
UNSPSC Code:
12352302
EC Index Number:
234-390-0
NACRES:
NA.21

Quality Level

Assay

≥96.0% (manganometric)

form

powder

potency

2567 mg/kg LD50, oral (Rat)

pH

10 (20 °C, 15 g/L in H2O)

mp

60 °C (decomposition)

density

1.73 g/cm3

bulk density

800 kg/m3

anion traces

chloride (Cl-): ≤0.1%
sulfate (SO42-): ≤1.2%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.003%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[B-]1(OO[B-](OO1)(O)O)(O)O.O.O.O.O.O.O

InChI

1S/B2H4O8.2Na.6H2O/c3-1(4)7-9-2(5,6)10-8-1;;;;;;;;/h3-6H;;;6*1H2/q-2;2*+1;;;;;;

InChI key

MCYPYXIRGFEFGL-UHFFFAOYSA-N

Linkage

Replaces: 1.06560.9057

Analysis Note

Assay (manganometric): ≥ 96.0 %
Identity: passes test
Chloride (Cl): ≤ 0.1 %
Sulfate (SO₄): ≤ 1.2 %
Heavy metals (as Pb): ≤ 0.003 %
Fe (Iron): ≤ 0.002 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service