Skip to Content
Merck
All Photos(2)

Key Documents

1.05056

SAFC

Potassium disulfite

E 224

Pharma Manufacturing

Synonym(s):

Potassium disulfite, Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2O5S2
CAS Number:
Molecular Weight:
222.32
MDL number:
UNSPSC Code:
12352302
E Number:
E224
EC Index Number:
240-795-3

Quality Level

form

solid

pH

3.0-4.5 (20 °C, 50 g/L in H2O)

mp

150 °C (decomposition)

application(s)

pharma/biopharma processes

storage temp.

15-25°C

SMILES string

[K+].[K+].[S](=O)(=O)([S](=O)[O-])[O-]

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Related Categories

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Potassium disulfite shows preservative and antioxidant properties. Potassium disulfite E224 is suitable for use in pharmaceutical processes and as food additive.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service