Skip to Content
Merck
All Photos(1)

Key Documents

1.04965

Supelco

Potassium cyanide

Potassium cyanide
1 of 1 reviewers received a sample product or took part in a promotion

EMPLURA®

Synonym(s):

Potassium cyanide, Cyanogen potassium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CKN
CAS Number:
Molecular Weight:
65.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
205-792-3
NACRES:
NA.21
Assay:
≥96% (silver nitrate titration)
form:
solid
Pricing and availability is not currently available.

Quality Level

product line

EMPLURA®

Assay

≥96% (silver nitrate titration)

form

solid

potency

7.49 mg/kg LD50, oral (Rat)
14.29 mg/kg LD50, skin (Rabbit)

pH

11-12 (20 °C, 20 g/L in H2O)

bp

1625 °C/1013 hPa

mp

634 °C

density

1.55 g/cm3 at 20 °C

bulk density

750 kg/m3

anion traces

sulfide (S2-): ≤0.001%

cation traces

Na: ≤1%

storage temp.

15-25°C

SMILES string

[K+].N#[C-]

InChI

1S/CN.K/c1-2;/q-1;+1

InChI key

NNFCIKHAZHQZJG-UHFFFAOYSA-N

Related Categories

Application


  • Supramolecular Gels Based on C(3)-Symmetric Amides: This study demonstrates the application of potassium cyanide in the development of supramolecular gels used for anion-sensing and dye removal from water. It highlights the role of potassium cyanide in enhancing the functionality of materials designed for environmental cleanup and monitoring, aligning with sustainable research practices in material science (Kuppadakkath G et al., 2024).

  • Binucleate Rhizoctonia induced tomato resistance against Rhizoctonia solani: This research explores the use of potassium cyanide in studying plant-pathogen interactions, particularly focusing on enhancing tomato plant resistance against fungal diseases. The application in agricultural biotechnology demonstrates the potential of potassium cyanide in developing disease-resistant crop varieties, contributing to sustainable agriculture practices (Taheri P et al., 2024).

Analysis Note

Assay (argentometric): ≥ 96 %
Sulphide (S): ≤ 0.001 %
Na (Sodium): ≤ 1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1.  Hello ,I have a question about one of your product (Potassium Cyanide )is it good for gold plating ?and is it powder?

    1 answer
    1. Product 901670, Bright electroless gold plating solution (https://www.sigmaaldrich.com/product/aldrich/901670), would be a more appropriate product to use for gold plating. It contains potassium dicyanoaurate(I) (or potassium gold cyanide) as the cyanide component, which is common in the plating industry.

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service