Skip to Content
Merck
All Photos(1)

Key Documents

102466-M

Sigma-Aldrich

Ethylene glycol

reagent grade, ≥99%

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12352137
eCl@ss:
39020501
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

752 °F

expl. lim.

15.3 %

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

solubility

water: miscible

density

1.113 g/mL at 25 °C (lit.)

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol (EG) is a polyol having a wide range of industrial applications. Its preparation from cellulose has been reported. It can also be produced from fossil fuels (petroleum, natural gas and coal) and biomass-derived resources.

Application

Ethylene glycol has been used as a carrier fluid during the fabrication of shear thickening fluid (composed of nanosized silica particles). It has also been used to compose previtrification and vitrification solutions for the cryopreservation of mouse embryos.
Reagent typically used in cyclocondensation reactions with aldehydes and ketones to form 1,3-dioxolanes.

Other Notes

Important notice
This product will be phased out towards early 2019. There is a replacement product available : 102466

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Study on magnetorheological shear thickening fluid.
Zhang X, et al.
Smart Materials and Structures, 17(1), 015051-015051 (2008)
Siloxanes as temporary tethers in [2+2] photocycloadditions
Crimmins MT and Guise LE
Tetrahedron Letters, 35(11), 1657-1660 (1994)
Sulfonated charcoal, a mild and efficient reagent for the preparation of cyclic acetals, dithioacetals and benzodioxepines
Patney HK
Tetrahedron Letters, 32(3), 413-416 (1991)
Aiqin Wang et al.
Accounts of chemical research, 46(7), 1377-1386 (2013-02-21)
With diminishing fossil resources and increasing concerns about environmental issues, searching for alternative fuels has gained interest in recent years. Cellulose, as the most abundant nonfood biomass on earth, is a promising renewable feedstock for production of fuels and chemicals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service