Skip to Content
Merck
All Photos(1)

Key Documents

I-055

Supelco

Isotonitazene hydrochloride solution

1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N4O3 · HCl
CAS Number:
Molecular Weight:
446.97
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol ((as free base))

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)OC(C=C1)=CC=C1CC2=NC3=C(N2CCN(CC)CC)C=CC([N+]([O-])=O)=C3.Cl

InChI

1S/C23H30N4O3.ClH/c1-5-25(6-2)13-14-26-22-12-9-19(27(28)29)16-21(22)24-23(26)15-18-7-10-20(11-8-18)30-17(3)4;/h7-12,16-17H,5-6,13-15H2,1-4H3;1H

InChI key

SPCSJKWVWNMACY-UHFFFAOYSA-N

Application


  • Quantitative analysis in forensic settings: Isotonitazene was included in a validation study of an analytical method for the quantitation of synthetic opioids in biological matrices, including plasma, blood, urine, liver, and brain, demonstrating its application in forensic toxicology and its role in authentic postmortem casework in urban settings (Pardi, Ford, and Cooper, 2023).

  • Opioid receptor binding affinity study: A non-radioactive mass spectrometry-based binding assay developed to measure the binding affinities of opioids, including Isotonitazene, emphasized its utility in pharmacological research, particularly for understanding interactions at the μ-opioid receptor, which is critical for developing new pain management therapies (Volz and Moosmann, 2022).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service