Skip to Content
Merck
All Photos(1)

Key Documents

867200O

Avanti

Squalene

from plant derived, sugarcane, Avanti Research - A Croda Brand

Synonym(s):

(6E,10E,14E,18E)-2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50
CAS Number:
Molecular Weight:
410.72
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

Assay

≥99% (TLC)

form

(neat oil)

packaging

pkg of 1 × 100 mg (867200O-100MG)
pkg of 5 × 100 mg (867200O-500MG)
pkg of 5 × 200 mg (67200O-1G)

manufacturer/tradename

Avanti Research - A Croda Brand

mp

-75 °C (lit)

application(s)

vaccine development

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

InChI

1S/C30H50/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h15-18,23-24H,9-14,19-22H2,1-8H3/b27-17+,28-18+,29-23+,30-24+

InChI key

YYGNTYWPHWGJRM-AAJYLUCBSA-N

General description

Squalene is a triterpene that is an intermediate in the cholesterol synthesis pathway . This product is a synthetic precursor to all steroids, including cholesterol, steroid hormones, and vitamin D.

Application

adjuvants

Packaging

5 mL Amber Glass Screw Cap Vial (867200O-100MG);5 mL Amber Glass Screw Cap Vial (867200O-500MG);5 mL Amber Glass Screw Cap Vial (867200O-1G)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service