Skip to Content
Merck
All Photos(2)

Key Documents

860453P

Avanti

16:0-d31-18:1 PA

1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

110920

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H39O8PNaD31
CAS Number:
Molecular Weight:
728.10
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860453P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860453P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C37H71O8P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(39)45-35(34-44-46(40,41)42)33-43-36(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,35H,3-16,19-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/b18-17-;/t35-;/m1./s1/i2D3,4D2,6D2,8D2,10D2,12D2,14D2,16D2,19D2,21D2,23D2,25D2,27D2,29D2,31D2;

InChI key

PPYXMAJBOKWTQX-VYSSPCEXSA-M

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-d31-18:1 PA has been used as an internal standard in yeast lipid profiling.

Packaging

5 mL Clear Glass Sealed Ampule (860453P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compartmentalized synthesis of triacylglycerol at the inner nuclear membrane regulates nuclear organization
Barbosa AD, et al.
Developmental Cell, 50(6), 755-766 (2019)
Jordon M Inloes et al.
Biochemistry, 57(39), 5759-5767 (2018-09-18)
Deleterious mutations in the serine hydrolase DDHD domain containing 1 (DDHD1) cause the SPG28 subtype of the neurological disease hereditary spastic paraplegia (HSP), which is characterized by axonal neuropathy and gait impairments. DDHD1 has been shown to display PLA1-type phospholipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service