Skip to Content
Merck
All Photos(2)

Key Documents

858143C

Avanti

18:1 Lyso PS

1-oleoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Synonym(s):

1-(9Z-octadecenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); PS(18:1(9Z)/0:0); 18:1 LPS; C18:1 LPS; 110726

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H45NO9PNa
CAS Number:
Molecular Weight:
545.58
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

99% (LPS; may contain up to 15% of the 2-LPS isomer, TLC)

form

liquid

packaging

pkg of 1 × 1 mL (858143C-10mg)
pkg of 1 × 1 mL (858143C-5mg)
pkg of 1 × 2.5 mL (858143C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 858143C

concentration

10 mg/mL (858143C-10mg)
10 mg/mL (858143C-25mg)
5 mg/mL (858143C-5mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)([H])COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI key

ULNYNKJABITZRB-UZADSWFSSA-M

Application

18:1 Lyso PS has been used as a lipid standard in the optimization of the liquid chromatography-mass spectrometry (LC-MS) method conditions.

Packaging

5 mL Clear Glass Sealed Ampule (858143C-10mg)
5 mL Clear Glass Sealed Ampule (858143C-25mg)
5 mL Clear Glass Sealed Ampule (858143C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingxin Shi et al.
Food & function, 10(12), 7782-7792 (2019-11-30)
Triterpenoid saponins from Kuding tea have demonstrated preventive effects on hyperlipidaemia induced by a high-fat diet. Lysoglycerophospholipids (Lyso-GPLs) are known to be associated with proatherogenic conditions such as hyperlipidaemia. In this study, a target profiling strategy based on a multiple
Wuhanqimuge et al.
FEBS open bio, 3, 243-251 (2013-06-19)
Lysophosphatidylcholine (LPC) is one of the major lysophospholipids mainly generated by phospholipase A2 (PLA2)-mediated hydrolysis of phosphatidylcholine (PC). We previously found that LPC displays neurotrophin-like activity in the rat pheochromocytoma PC12 cells and in cerebellar granule neurons, but the molecular
Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate
Jung Yong Eum et al.
Journal of chromatography. A, 460849-460849 (2020-01-14)
Aging refers to the intracellular accumulation of reactive oxygen species that damages proteins, DNA, and lipids. As alterations in lipid metabolism may trigger metabolic disorders and the onset of metabolic diseases, changes in lipid profiles can be closely related to
Masako Nishikawa et al.
Journal of atherosclerosis and thrombosis, 22(5), 518-526 (2014-12-03)
Lysophospholipids, particularly sphingosine 1-phosphate and lysophosphatidic acid, are known to be involved in the pathogenesis of atherosclerosis; however, the role of lysophosphatidylserine (LysoPS) in the onset of atherosclerotic diseases remains uncertain. We investigated the effects of LysoPS on the uptake

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service