Skip to Content
Merck
All Photos(1)

Key Documents

330731

Avanti

EquiSPLASH

Avanti Research - A Croda Brand

Synonym(s):

EQUISPLASH LIPIDOMIX QUANTITATIVE MASS SPEC INTERNAL STANDARD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.28

form

methanol solution

packaging

pkg of 1 × 1 ea (330731-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

General description

Single-vial Prepared Lipidomic Analytical Standard

Mixture of 13 deuterated lipid internal standards at a concentration of 100μg/mL each.  Each sealed ampule contains 1mL methanol solution. Concentrations are verified for use as a quantitative standard.

Mixture Component
  • 15:0-18:1(d7) PC
  • 18:1(d7) Lyso PC
  • 15:0-18:1(d7) PE
  • 18:1(d7) Lyso PE
  • 15:0-18:1(d7) PG (Na Salt)
  • 15:0-18:1(d7) PI (NH4 Salt)
  • 15:0-18:1(d7) PS (Na Salt)
  • 15:0-18:1(d7)-15:0 TAG
  • 15:0-18:1(d7) DAG
  • 18:1(d7) MAG
  • 18:1(d7) Chol Ester
  • d18:1-18:1(d9) SM
  • C15 Ceramide-d7

Application

EquiSPLASH is suitable for use as internal standard for matrix assisted laser desorption/ionization-time of flight (MALDI-TOF) analysis.

Packaging

2ML Amber Glass Sealed Ampule (330731-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
EquiSPLASH is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 791637C15:0-18:1-d7-PC, Avanti Research - A Croda Brand 100μg/mLSDS

  • 791643C18:1-d7 Lyso PC, Avanti Research - A Croda Brand 100μg/mLSDS

  • 791638C15:0-18:1-d7-PE, Avanti Research - A Croda Brand 100μg/mLSDS

  • 791644C18:1-d7 Lyso PE, Avanti Research - A Croda Brand 791644C 100μg/mLSDS

  • 791640C15:0-18:1-d7-PG, Avanti Research - A Croda Brand 791640C 100μg/mLSDS

  • 791641C15:0-18:1-d7-PI, Avanti Research - A Croda Brand 100μg/mLSDS

  • 791639C15:0-18:1-d7-PS, Avanti Research - A Croda Brand 100μg/mLSDS

  • 791648C15:0-18:1-d7-15:0 TG, Avanti Research - A Croda Brand 791648C 100μg/mLSDS

  • 15:0-18:1-d7-15:0 TG 100μg/mL

  • 791646C18:1-d7 MG, Avanti Research - A Croda Brand 791646C 100μg/mLSDS

  • 791645C18:1-d7-cholesterol, Avanti Research - A Croda Brand 791645C 100μg/mLSDS

  • 791649C18:1-d9 SM, Avanti Research - A Croda Brand 100μg/mLSDS

  • 860681PC15 Ceramide-d7 (d18:1-d7/15:0), Avanti Research - A Croda Brand 860681P, powder 100μg/mLSDS

See All (13)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative evaluation of the extraction and analysis of urinary phospholipids and lysophospholipids using MALDI-TOF/MS
Li X, et al.
Chemistry and Physics of Lipids, 223 (2019)
Akira Utsunomiya et al.
The Journal of investigative dermatology (2019-11-02)
Dermokine is a chiefly skin-specific secreted glycoprotein localized in the upper epidermis, and its family consists of three splice variants in mice and five in humans. To investigate the pathophysiological impact of dermokine, we generated mice deficient for two (βγ)
Lipidomics Profiling Reveals Differential Alterations after FAS Inhibition in 3D Colon Cancer Cell Culture Models
Fries B D, et al.
Journal of Proteome Research (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service